Anti-LMO4 Rabbit Monoclonal Antibody [clone: D6V4Z]
CSTA81428S
- Type antilichaam:Primair
- antigeen symbool:LMO4
- Clonality:Monoklonaal
- Clone:D6V4Z
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Sequentie:CRNRLVPGDRFHYINGSLF
- Opslagtemperatuur:–20 °C
- Shipping temperature:Ambient
- VPE:100 µl
Type: Primary
Antigen: LMO4
Clonality: Monoclonal
Clone: D6V4Z
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse