Anti-FUNDC1 Rabbit Monoclonal Antibody [clone: E1C6G]
Catalogus # CSTA97655S
Leverancier: Cell Signaling
Specificaties
- Type antilichaam:Primair
- antigeen symbool:FUNDC1
- Clonality:Monoklonaal
- Clone:E1C6G
- Host:Rabbit
- ImmunoNeerslag:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequentie:ATRNPPPQDYESDDDSYEVLDLTEYARRHQC
- Opslagtemperatuur:−20 °C
- Shipping temperature:Cold
- Maat:100 µl
- VPE:100 µl
Specificaties
Over dit artikel
Type: Primary Antibodies
Antigen: FUNDC1
Clonality: Monoclonal
Clone: E1C6G
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human