Anti-NUAK2/SNARK Rabbit Monoclonal Antibody [clone: E4V4Q]
Catalog # CSTA15452S
Supplier: Cell Signaling
Specifications
- Type antilichaam:Primair
- antigeen symbool:NUAK2/SNARK
- Clonality:Monoklonaal
- Clone:E4V4Q
- Host:Rabbit
- ImmunoNeerslag:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequentie:Q9H093 L472-T628
- Opslagtemperatuur:−20 °C
- Shipping temperature:Ambient
- Maat:100 µl
- VPE:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: NUAK2/SNARK
Clonality: Monoclonal
Clone: E4V4Q
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat