Anti-ARD1A Rabbit Polyclonal Antibody
Catalog # CSTA9046S
Supplier: Cell Signaling
Specifications
- Type antilichaam:Primair
- antigeen symbool:ARD1A
- Clonality:Polyklonaal
- Host:Rabbit
- ImmunoNeerslag:Yes
- Reactivity:Monkey,Human
- Western blot:Yes
- Sequentie:CRHLELKEKGRHVVLGAIENK
- Opslagtemperatuur:−20 °C
- Shipping temperature:Cold
- Maat:100 µl
- VPE:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: ARD1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Human,Monkey