Anti-RAD54 Rabbit Monoclonal Antibody [clone: D4W3Z]
CSTA15016S
- Type antilichaam:Primair
- antigeen symbool:RAD54
- Clonality:Monoklonaal
- Clone:D4W3Z
- Host:Rabbit
- ImmunoNeerslag:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequentie:CIDGGSKDEIDQKLEG
- Opslagtemperatuur:−20 °C
- Shipping temperature:Ambient
- Maat:100 µl
- VPE:100 µl
Type: Primary Antibodies
Antigen: RAD54
Clonality: Monoclonal
Clone: D4W3Z
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human