Anti-MAGE-C2 Rabbit Monoclonal Antibody [clone: E7B8D]
Type: Primary
Antigen: MAGE-C2
Clonality: Monoclonal
Clone: E7B8D
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- CSTA20058S
- CSTA20058T
- antigeen symbool:
- MAGE-C2
- MAGE-C2
- Type antilichaam:
- Primair
- Primair
- Clonality:
- Monoklonaal
- Monoklonaal
- Clone:
- E7B8D
- E7B8D
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemie:
- Yes
- Yes
- ImmunoNeerslag:
- Yes
- Yes
- Sequentie:
- CEEVPSGVIPNLTESIPSS
- CEEVPSGVIPNLTESIPSS
- Opslagtemperatuur:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Frequently Bought Together