Orders LinkContactUsLinkComponent
PELTOR™ LiteCom Plus Headset
PELTOR™ LiteCom Plus Headset
PELTOR™ LiteCom Plus Headset, 32 dB
Catalogus # MMMAMT73H7A4410EU
Leverancier:   3M
PELTOR™ LiteCom Plus Headset
Catalogus # MMMAMT73H7A4410EU
Leverancier:  3M
PELTOR™ LiteCom Plus Headset
PELTOR™ LiteCom Plus Headset, 32 dB
Price
1 540,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
3M™ PELTOR™ LiteCom Plus Headset is our robust hearing protectors with a built-in analogue two-way communication radio, noise cancelling dynamic microphone, Lithium-Ion battery and level dependent functionality for ambient listening. Available in headband and helmet mounted models.
  • Noise cancelling dynamic microphone provides voice-operated transmission for hands free use
  • External connection jack allows connection of a mobile device or another two-way radio
  • Lithium-Ion battery
  • Auto power off and clear battery status indication
  • Available in headband and helmet mounted models
  • Available in 16 channel PMR 446 MHz version with 121 sub-channels
3M™ PELTOR™ LiteCom Plus Headset feature a built-in analogue two-way radio, available in PMR version with 16 channels and 121 sub-channels - to help you communicate even more efficiently in noisy environments. The noise cancelling dynamic microphone provides voice-operated transmission (VOX) for hands free use, and an external jack allows you to connect a mobile device or another two-way radio. Sound levels constantly change in demanding situations.
When the noise quiets down, you can turn up the environmental listening function to improve situational awareness or speak with co-workers who are close by, and if the hazardous noise returns the level dependent technology will kick in to help protect your hearing again. Litecom Plus Headsets have a clear battery status indicator and an auto power off function to save energy.
Specificaties
  • Hoogte legplaat:
    32 dB
  • Omschrijving:
    PELTOR™ LiteCom Plus Headset