Amantadine
Glass bottle for solids, 25 g
MOLEM61882414
Marketsource
: Molekula
: 1-Adamantylamine, 1-Aminotricyclo[3.3.1.1(3,7)]decane, Adamantan-1-amine, 1-Adamantanamine, 1-Aminoadamantane
- :768-94-5
- :C₁₀H₁₇N
- :MFCD00074732
- :151.25 g/mol
- :12,00389
- :214 °C (1013 hPa)
- :82 °C
- :212-201-2
- Verpakking:Glass bottle for solids
- VPE:25 g