Furfurylalcohol
Catalog # EHERC13972300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Verpakking:Glass bottle for solids
- VPE:0,25 g
- Seq:3029
Specifications
About this item
:
:98.1 g/mol
Merck Index:13,04326
Boiling Pt:171 °C (1013 hPa)
Melting Pt:–31 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:75 °C (closed cup)
Storage Temperature:Kamertemperatuur
EINECS:202-626-1
UN:2874
ADR:6.1,III
:GHS06
:GHS08
CAS Number:98-00-0
Formula:C₅H₆O₂
MDL Number:MFCD00003252