Digestive system
Digestive system, K21
Catalogus # 763-0125
Marketsource
Leverancier: 3B Scientific
Ga naar:
Product Details & Documents
Life-size model that shows the entire digestive system in relief: Nose, mouth, throat, etc. via the intestinal package. Dimensions 81×33×10 cm.
Specificaties
- Omschrijving:Digestive system, K21