Nerve cell neuron
Illustration of nerves, Metaal
Catalog # 763-0107
Marketsource
Supplier: 3B Scientific
Jump to:
Product Details & Documents
Base plate with cut surface of a nerve and a model in 5 different parts that are mounted with magnets. Shows the structure of the nerve cell and its connections with the muscle cells. Instructions for use included.
- Dimensions 68 × 51 × 3 cm
Specifications
- Omschrijving:Illustration of nerves
- Materiaal:Metaal