Order Entry
Belgium
ContactUsLinkComponent
8-Bromo-6-chloroimidazo[1,2-a]pyridine 95%
Irritant
8-Bromo-6-chloroimidazo[1,2-a]pyridine 95%
Catalog # APOSOR110712-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
8-Bromo-6-chloroimidazo[1,2-a]pyridine 95%
Catalog # APOSOR110712-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Verpakking:
    Bottle
  • VPE:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :231.48 g/mol
 :GHS07
CAS Number:957187-27-8
Formula:C₇H₄BrClN₂
MDL Number:MFCD12828062