Tetracysteine FLASH Diacetate *2 mM DMSO Solution*
Catalog # AATB22334
Supplier: AAT Bioquest
CAS Number:
Specifications
- VPE:100 Tests
- Omschrijving:Tetracysteine FLASH diacetate *2 mM DMSO solution*
- Tests per kit:100 Tests
Specifications
About this item
FLASH diacetate is a cell-permeable version of FlASH, which is a fluorescein derivative, modified to contain two arsenic atoms at a set distance from each other.
- Fluorescein derivative
- High affinity for thiols
- Multiple applications
Caution: For research use only.