ReadiLink™ Rapid trFluor™ Eu Antibody Labeling Kit "Microscale Optimised for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalogus # AATB1300
Leverancier: AAT Bioquest
CAS-nummer:
Specificaties
- VPE:2 Tests
- Omschrijving:ReadiLink™ rapid trFluor™ Eu antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Specificaties
Over dit artikel
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- Easy and simple protocol for Eu labeling
- TRF assay development
- Large Stokes shift
Let op: For research use only.