Protonex™ Green 500 Dextran
1 mg
AATB21217
Protonex™ Green dye demonstrated pH-dependent fluorescence.
- pH-dependent fluorescence
- No fluorescence outside the cell
- Excellent for imaging acidic organelles
- Cell impermeable
Caution: For research use only.
- VPE:1 mg