Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Protonex™ Green 500 Dextran
1 mg
  AATB21217
Protonex™ Green 500 Dextran
  AATB21217
 :  
Protonex™ Green 500 Dextran
Protonex™ Green 500 Dextran
1 mg
 :  
313,00 €
1 * 1 mg
Protonex™ Green dye demonstrated pH-dependent fluorescence.
  • pH-dependent fluorescence
  • No fluorescence outside the cell
  • Excellent for imaging acidic organelles
  • Cell impermeable
Caution: For research use only.
  • VPE:
    1 mg