Orders LinkContactUsLinkComponent
Cell Explorer™ Live Cell Tracking Kit
Cell Explorer™ Live Cell Tracking Kit
Kit is designed to uniformly label live cells in blue fluorescence for the studies that require the fluorescent tag molecules retained inside cells for relatively longer time, 200 Tests
Catalogus # AATB22620
Leverancier:   AAT Bioquest
Cell Explorer™ Live Cell Tracking Kit
Catalogus # AATB22620
Leverancier:  AAT Bioquest
CAS-nummer:  
Cell Explorer™ Live Cell Tracking Kit
Cell Explorer™ Live Cell Tracking Kit
Kit is designed to uniformly label live cells in blue fluorescence for the studies that require the fluorescent tag molecules retained inside cells for relatively longer time, 200 Tests
CAS-nummer:  
Price
313,00 €
Log in voor beschikbaarheid
1 * 200 Tests
Product Details & Documents
The kit uses a weakly fluorescent dye that carries a cell-retaining moiety.
  • Long term tracking
  • Suitable for variety of studies
  • Optimized cell labeling protocol
The labeling process is robust, requiring minimal hands-on time. It can be readily adapted for a wide variety of fluorescence platforms such as microplate assays, immunocytochemistry and flow cytometry.
Verpakking: Plastic vial.
Let op: For research use only.
Specificaties
Specificaties
  • VPE:
    200 Tests
  • Omschrijving:
    Kit is designed to uniformly label live cells in blue fluorescence for the studies that require the fluorescent tag molecules retained inside cells for relatively longer time