ReadiLink™ Rapid mFluor™ Red 700 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1130
Supplier: AAT Bioquest
CAS Number:
Specifications
- VPE:2 Tests
- Omschrijving:ReadiLink™ rapid mFluor™ red 700 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
Specifications
About this item
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
- Red-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.