ReadiLink™ Rapid iFluor® 350 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1220
Supplier: AAT Bioquest
CAS Number:
Specifications
- VPE:2 Tests
- Omschrijving:ReadiLink™ rapid iFluor™ 350 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
Specifications
About this item
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
- Violet-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.