Tetracysteine REASH Acetate *2 mM DMSO Solution*
Catalogus # AATB22336
Leverancier: AAT Bioquest
CAS-nummer:
Specificaties
- VPE:100 Tests
- Omschrijving:Tetracysteine REASH acetate *2 mM DMSO solution*
- Tests per kit:100 Tests
Specificaties
Over dit artikel
REASH acetate is a cell-permeable version of REASH, which is a resorufin derivative, modified to contain two arsenic atoms at a set distance from each other.
- Fluorescein derivative
- High affinity for thiols
- Multiple applications
Let op: For research use only.