Tetracysteine FLASH Reagent *2 mM DMSO Solution*
Catalogus # AATB22330
Leverancier: AAT Bioquest
CAS-nummer:
Specificaties
- VPE:100 Tests
- Omschrijving:Tetracysteine FLASH reagent *2 mM DMSO solution*
- Tests per kit:100 Tests
Specificaties
Over dit artikel
FLASH is a fluorescein derivative, modified to contain two arsenic atoms at a set distance from each other.
- Fluorescein derivative
- High affinity for thiols
- Multiple applications
Let op: For research use only.