Cell Navigator® Mitochondrion Staining Kit *Deep Red Fluorescence*
Catalog # AATB22669
Supplier: AAT Bioquest
CAS Number:
Specifications
- VPE:500 Assays
- Omschrijving:Cell Navigator™ Mitochondrion staining kit *Deep red fluorescence*
Specifications
About this item
The Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labeling sub-cellular organelles such as membranes, lysosomes, mitochondria and nuclei etc.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Caution: For research use only.