Clips for conical standard ground joints
Catalogus # BRND55646
Leverancier: Brand
Specificaties
- Voor NS:29/32
- Kleur:rood
- VPE:1
Specificaties
Over dit artikel
POM. Will not scratch glass surfaces.
- Excellent resilience and stability
- Outstanding chemical resistance
- Operating temperature up to 150 °C