Orders LinkContactUsLinkComponent
Adapters, Cone without Stopcock
Adapters, Cone without Stopcock
, 29/32 NS, Slangen
Catalogus # WITG1.681.029
Leverancier:   witeg Labortechnik
Adapters, Cone without Stopcock
Catalogus # WITG1.681.029
Leverancier:  witeg Labortechnik
Adapters, Cone without Stopcock
, 29/32 NS, Slangen
Price
16,30 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Suction tube, with cone and without stopcock, bent.
Specificaties
  • Afmetingen kern:
    29/32 NS
  • Seq:
    05
  • Slangverbinding:
    Slangen