Orders LinkContactUsLinkComponent
Experimentation kits, size exclusion chromatography
Size Exclusion Chromatography
Catalog # BE-414
Supplier:   G-Biosciences
Experimentation kits, size exclusion chromatography
Catalog # BE-414
Supplier:  G-Biosciences
Experimentation kits, size exclusion chromatography
Experimentation kits, size exclusion chromatography
Size Exclusion Chromatography
Price
166,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The Size Exclusion Chromatography kit teaches gel filtration or size exclusion chromatography and the use of this method in purification of biological samples. This method is based on separation of protein molecules based on their molecular size. This lab activity involves running size exclusion chromatography for separation of molecules based on their molecular sizes.
  • Principles of gel filtration or size exclusion chromatography
  • Factors affecting gel filtration chromatography
  • Hands-on chromatography lab activity
  • Role of gel filtration and strategies in protein purification
Also required: Clamp and stand.
Delivery information: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Documents
Specifications
  • Omschrijving:
    Size Exclusion Chromatography