Experimentation kits, hydrophobic chromatography
Catalog # BE-416
Supplier: G-Biosciences
Specifications
- Omschrijving:Hydrophobic Chromatography
- VPE:1 KIT
Specifications
About this item
The Hydrophobic Chromatography kit is designed to teach students the basic principle of hydrophobic chromatography utilising a hydrophobic enzyme. The use of the enzyme allows purification followed by a simple enzyme assay to detect the fractions that contain the enzyme. This lab activity involves preparation of a crude protein extract and running hydrophobic chromatography to isolate the enzyme.
- The principle of hydrophobic chromatography
- Immobilisation of protein on hydrophobic columns
- Factors influencing binding and elution of proteins on hydrophobic columns
- Hands-on hydrophobic chromatography lab activity
Also required: Low speed centrifuge for 1,5 to 2 ml tubes, water bath or beaker and thermometer.
Delivery: Supplied with components needed for hands-on experimentation for six workstations of 4-5 students or 24-30 students. Supplied with Teacher’s Guide and separate Student’s Guides.