Do Onions, Strawberries and Bananas Have DNA?
Catalog # EDVOS-75
Supplier: EDVOTEK
Specifications
- Omschrijving:Do Onions, Strawberries and Bananas Have DNA?
- VPE:1
Specifications
About this item
Kit allows students to construct DNA models and then extract DNA from onions, strawberries or bananas.
- For 10 lab groups
- Complete in 30 minutes
Also required: 95 to 100% isopropyl alcohol, fruit and vegetables.
Also required: 95-100% isopropyl alcohol, fruit and vegetables.
Delivery: Kit includes instructions, DNA extraction buffer, DNA sample in capped test tube, transfer pipettes, pop beads, glass rod, DNA spooling rods, test tubes, and salt.