Genes in a Tube™
Catalogus # EDVO119
Leverancier: EDVOTEK
Specificaties
- Omschrijving:Genes in a Tube™
- VPE:1
Specificaties
Over dit artikel
Kit for teaching students how to extract and precipitate their own DNA. Students can transfer their DNA to a tube that can be used as a pendant on a necklace.
- For 25 students, plus 1
- Complete in 30 minutes
Also required: Ice cold alcohol (95% ethanol or isopropyl alcohol) and a 56 °C water bath.
Also required: Ice cold alcohol (95% ethanol or isopropyl alcohol) and a 56 °C water bath.
Levering: Kit includes instructions, lysis buffer, NaCl solution, Protease, Tris buffer, Methylene Blue Plus solution, microcentrifuge tubes, sterile cotton tipped applicators, transfer pipettes, tubes for DNA precipitation, Gene Tubes, and string.