Orders LinkContactUsLinkComponent
Mini-Pellet Press Basic Solid Pack
Mini-Pellet Press Basic Solid Pack
Mini pellet press kit
Catalog # SPCCGS01150
Supplier:   SPECAC
Mini-Pellet Press Basic Solid Pack
Catalog # SPCCGS01150
Supplier:  SPECAC
Mini-Pellet Press Basic Solid Pack
Mini-Pellet Press Basic Solid Pack
Mini pellet press kit
Price
4 320,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Sampling packs provide everything you need to get started measuring the FTIR spectra of solids, liquids or gases. Starter kits bundle together the sampling packs for a cost effective way to maximise your labs versatility.
  • Full hydraulic operation and integrated pressure gauge
  • Simple operation requiring minimal training
  • Small and lightweight
  • Low cost of ownership
  • Ideal for FTIR/KBr pellet preparation
The Specac Mini Pellet Press is an entry-level, handheld laboratory hydraulic press designed for KBr sample preparation. It offers quick and efficient preparation of high-quality 7 mm KBr pellets for transmission FTIR analysis, utilizing up to 2 tons of pressure.
With its rugged and durable design, this portable mini hydraulic press is intended for frequent use in the laboratory, offering maintenance-free operation.
Delivery information: Includes a mini-pellet press and die set and KBr powder, 7 mm pellet Die including pellet ring holder, 7 mm spare pellet ring holder, 7 mm disc holder with rectangular mount, pestle and mortar, 50 g KBr Power.
Specifications
  • Omschrijving:
    Mini pellet press kit
  • Seq:
    01