Orders LinkContactUsLinkComponent
Particulate Filter, DT
Particulate Filter, DT
Particulate filter, PF10 P3 R D
Catalog # MMMADT-1135E
Supplier:   3M
Particulate Filter, DT
Catalog # MMMADT-1135E
Supplier:  3M
Particulate Filter, DT
Particulate Filter, DT
Particulate filter, PF10 P3 R D
Price
152,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
For effective protection against a range of particulates, gases and vapours or combination hazards, use 3M™ filters DT-Series. Suitable for use with certain 3M™ half and full-face tight-fitting respirators in negative-pressure mode as well as 3M™ powered air respirator system PF-600E+ series.
  • Compatible with certain supplied air respiratory systems
  • Features a DIN40 mm threaded connection
3M™ filters DT-Series are available in a range of specifications to provide protection against particulate hazards, gas and vapour hazards or a combination of both. They are designed for use with 3M reusable and powered air respirators with DIN 40 filter ports.
Recommended application: Demolition, grinding, metalworking
Specifications
  • Omschrijving:
    Particulate filter, PF10 P3 R D