Sentinel XE® Complete PAPR System with Hood
Catalogus # ILCDS-3500EU
Leverancier: ILC DOVER
Specificaties
- Omschrijving:Sentinel XE® complete PAPR system with hood, CE marked
- Seq:01
- VPE:1
Specificaties
Over dit artikel
Sentinel XE® PAPR (powered air-purifying respirator) with unique clear hood is specifically designed to delivery TH3 level protection to pharmaceutical and biopharmaceutical workers.
- Improved safety with a 320° field of view panoramic hood
- Better comfort; no ratcheted head harness
- Greater airflow capacity delivers 220 LPM airflow within the hood and suit
- Lightweight blower provides better ergonomics
- Longer battery life; 12+ hours of service increases productivity
The Sentinel XE® complete PAPR system with hood is available with CE or UKCA marking.
Certificeringen: Approved to EN 12941/A2:208 TH3 P R SL. TH3 is the highest level of production and performance available under this certification. RoHS compliant per 2011/65/EU.