Over dit artikel
Micro-Touch® NitraFree™ examination gloves are scientifically designed to complement every need ofmedical professionals. Sophisticated design features include Ansell ergonomic technologies that ensure comfort during extended procedures, wet grip innovations that prevent harmful mishandling and micro-thin films for maximum sensitivity. Caregivers who are fully prepared and comfortably protected by versatile Micro-Touch® styles have the peace of mind to focus on what matters most: the health and protection of themselves and their patients.
- Reduced allergy risk
- Comfortable fit
- Heightened tactility
- Specialized protection
Micro-Touch® NitraFree™ examination glove are tested for use with chemotherapy drugs, standard patient examinations, protection from latex Type I allergy and minimise chemical Type IV allergies in HCW's or patients.
Manufactured without sulfur-based chemical accelerators, helping to protect users from Type IV chemical allergies.
Keep out of direct sunlight, store in a cool and dry place. Keep away from sources of ozone or ignition.
Specificaties
- Catalog No:
- ANSM6034514
- ANSM6034513
- ANSM6034512
- ANSM6034511
- ANSM6034510
- Voor eenmalig gebruik/herbruikbaar:
- Voor eenmalig gebruik
- Voor eenmalig gebruik
- Voor eenmalig gebruik
- Voor eenmalig gebruik
- Voor eenmalig gebruik
- Latex/latexvrij:
- Latexvrij
- Latexvrij
- Latexvrij
- Latexvrij
- Latexvrij
- Gepoederd/poedervrij:
- poedervrij
- poedervrij
- poedervrij
- poedervrij
- poedervrij
- Materiaal:
- Nitril
- Nitril
- Nitril
- Nitril
- Nitril
- Afwerking:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Manchetstijl:
- Rolrand
- Rolrand
- Rolrand
- Rolrand
- Rolrand
- Vingerdikte:
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- Handpalmdikte:
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- FDA gereguleerd:
- +
- +
- +
- +
- +
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- ASTM-nr.:
- D6319
- D6319
- D6319
- D6319
- D6319