Orders LinkContactUsLinkComponent
TAAG E41 Universal Pathogens
TAAG E41 Universal Pathogens
TAAG E41 universal pathogens, V-FE04-1, 1 jar - 500 g, 1
  TAAGTGG0003
TAAG E41 Universal Pathogens
  TAAGTGG0003
 :  
TAAG E41 Universal Pathogens
TAAG E41 Universal Pathogens
TAAG E41 universal pathogens, V-FE04-1, 1 jar - 500 g, 1
 :  
535,00 €
1 * 1 ST
Dehydrated enrichment media for growing Salmonella spp., Staphylococcus aureus, Escherichia coli and Listeria monocytogenes in food samples. It contains selected growth factors and nutritional requirements optimized to promote pathogens development.
  • Model:
    V-FE04-1
  • VPE:
    1
  • Omschrijving:
    TAAG E41 universal pathogens
  • Bestanddelen:
    1 jar - 500 g