Orders LinkContactUsLinkComponent
TAAG E31 Universal Surfaces
TAAG E31 Universal Surfaces
TAAG E31 universal surfaces, V-FF03-1, 1 jar - 500 g, 1
Catalog # TAAGTGG0002
Supplier:   KREGLINGER EUROPE
TAAG E31 Universal Surfaces
Catalog # TAAGTGG0002
Supplier:  KREGLINGER EUROPE
CAS Number:  
TAAG E31 Universal Surfaces
TAAG E31 Universal Surfaces
TAAG E31 universal surfaces, V-FF03-1, 1 jar - 500 g, 1
CAS Number:  
Price
647,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dehydrated enrichment media for growing Salmonella spp., Staphylococcus aureus, Escherichia coli and Listeria monocytogenes in surface samples. It contains selected growth factors and nutritional requirements optimized to promote pathogens development.
Specifications
Specifications
  • Model:
    V-FF03-1
  • VPE:
    1
  • Omschrijving:
    TAAG E31 universal surfaces
  • Bestanddelen:
    1 jar - 500 g
Specifications