Polka Dot Plus Gloves
Ga naar:
Over dit artikel
PVC dots on both sides extend the life of this glove. Perfect for warehousing and distribution, these ambidextrous gloves can be worn on either hand.
- PVC dotted palm for enhanced grip
- Ambidextrous design suitable for left- and right-handed users
- Seamless 13 gauge liner
- Performs well in dry conditions
Low linting construction for minimal contamination, with a breathable seamless liner.
Colour: White/blue
Certificeringen: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 1142X. CE certified.
2 Opties Hieronder Beschikbaar
Product Details & Documents
PVC dots on both sides extend the life of this glove. Perfect for warehousing and distribution, these ambidextrous gloves can be worn on either hand.
- PVC dotted palm for enhanced grip
- Ambidextrous design suitable for left- and right-handed users
- Seamless 13 gauge liner
- Performs well in dry conditions
Low linting construction for minimal contamination, with a breathable seamless liner.
Colour: White/blue
Certificeringen: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 1142X. CE certified.
Documents
TDS A113 - Polka Dot Plus Gloves