Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Arctic Winter Gloves
M, geel
Catalogus # PRTWA146YERM
Leverancier:   Portwest
Arctic Winter Gloves
Catalogus # PRTWA146YERM
Leverancier:  Portwest
Arctic Winter Gloves
Arctic Winter Gloves
M, geel
Price
8,60 €
Log in voor beschikbaarheid
1 * 1 PAIRE
Product Details & Documents
Twin liner traps in heat, and a ³/₄ nitrile sandy dipping offers greater protection to the fingers as well as improved liquid protection. Open back for breathability. A fantastic glove for a wide range of industries.
  • Specially designed for use in cold conditions
  • Twin liner traps in heat through increased insulation
  • Provides contact heat protection up to 250 °C for 15 seconds
  • Flexible sandy nitrile coating offers great grip in wet and dry conditions
  • ³/₄ dipped for increased protection
  • Superb abrasion and tear resistance
  • 15 gauge liner for extra dexterity
  • OEKO-TEX® STANDARD 100 approved
Low linting construction for minimal contamination, with a breathable seamless liner. Retail tag aids presentation for retail sales.
Colour: Yellow
 : EN ISO 21420: 2020 Dexterity 3, EN 388: 2016 + A1: 2018 4242X, EN 407: 2020 X2XXXX, EN 511: 2006 X2X. ANSI/ISEA 105: 2016 CUT Level A7, ANSI/ISEA 105: 2016 ABRASION Level 4. CE certified.
Specificaties
  • Handschoenmaat:
    M
  • Kleur:
    geel
  • Gepoederd/poedervrij:
    poedervrij
  • Cat. No.:
    PRTWA146YERM
  • Voor eenmalig gebruik/herbruikbaar:
    Herbruikbaar
  • Latex/latexvrij:
    Latexvrij
  • Ambidexter/handspecifiek:
    Handspecifiek
  • Materiaal:
    Acryl/polyester
  • Sterility:
    Non sterile
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.