Arctic Winter Gloves
Product Details & Documents
Twin liner traps in heat, and a ³/₄ nitrile sandy dipping offers greater protection to the fingers as well as improved liquid protection. Open back for breathability. A fantastic glove for a wide range of industries.
- Specially designed for use in cold conditions
- Twin liner traps in heat through increased insulation
- Provides contact heat protection up to 250 °C for 15 seconds
- Flexible sandy nitrile coating offers great grip in wet and dry conditions
- ³/₄ dipped for increased protection
- Superb abrasion and tear resistance
- 15 gauge liner for extra dexterity
- OEKO-TEX® STANDARD 100 approved
Low linting construction for minimal contamination, with a breathable seamless liner. Retail tag aids presentation for retail sales.
Colour: Yellow
: EN ISO 21420: 2020 Dexterity 3, EN 388: 2016 + A1: 2018 4242X, EN 407: 2020 X2XXXX, EN 511: 2006 X2X. ANSI/ISEA 105: 2016 CUT Level A7, ANSI/ISEA 105: 2016 ABRASION Level 4. CE certified.
Specificaties
- Handschoenmaat:M
- Kleur:geel
- Gepoederd/poedervrij:poedervrij
- Cat. No.:PRTWA146YERM
- Voor eenmalig gebruik/herbruikbaar:Herbruikbaar
- Latex/latexvrij:Latexvrij
- Ambidexter/handspecifiek:Handspecifiek
- Materiaal:Acryl/polyester
- Sterility:Non sterile