Orders LinkContactUsLinkComponent
PYROSTAR™ Neo+ Endotoxin Detection Reagent
PYROSTAR™ Neo+ Endotoxin Detection Reagent
1 recombinant multi-test vial (2,7 ml), Sensitivity;: 0,001 EU/ml, 2,7 mL
Catalog # 293-36941
PYROSTAR™ Neo+ Endotoxin Detection Reagent
Catalog # 293-36941
CAS Number:  
PYROSTAR™ Neo+ Endotoxin Detection Reagent
PYROSTAR™ Neo+ Endotoxin Detection Reagent
1 recombinant multi-test vial (2,7 ml), Sensitivity;: 0,001 EU/ml, 2,7 mL
CAS Number:  
Price
437,00 €
Log in for availability
1 * 2,7 mL
Product Details & Documents
PYROSTAR Neo+ is a new endotoxin detection reagent developed by recombinant technology.
  • Colorimetric method, can be used with absorption plate reader
  • Same cascade reaction as Lysate reagent, conforms to traditional detection
  • Endotoxin specific reagent, no false positive risk of (1→3)-β-D-glucan
  • Determination range from 0,001 to 50 EU/ml
  • High sensitivity, reagent reacts with endotoxin over a wide range
Combine with traditional lysate reagent and recombinant 3 factors (factor C, factor B, Proclotting Enzyme) from Limilus polyphemus. It is a high sensitive colorimetric assay specific for endotoxin.
Specifications
Specifications
  • VPE:
    2,7 mL
  • Omschrijving:
    1 recombinant multi-test vial (2,7 ml), Sensitivity;: 0,001 EU/ml
Specifications