Welding Headbands and Headband Components
Catalog # MMMAH536200
Supplier: 3M
Specifications
- Omschrijving:Speedglas™ Welding helmet headband, back part, ratchet, 9100
- VPE:1
Specifications
About this item
Replacement head suspension assemblies, headbands and spare parts for 3M™ Speedglas™ Welding helmets.
- Highly adjustable and comfortable with a smooth ratchet for precise tightening
- Designed to be used with 3M™ Speedglas™ Welding Helmets
- Includes assembly parts
- For highest possible safety and comfort, regularly check your equipment for worn or damaged parts
Replacement suspension assemblies, headbands and spare parts for 3M™ Speedglas™ Welding helmets. Headband assemblies are highly adjustable and comfortable with a smooth ratchet for precise tightening. For maximum safety and comfort, regularly check your equipment for worn or damaged parts.