Orders LinkContactUsLinkComponent
Peltor™ Z3AF Back Plate
Peltor™ Z3AF Back Plate
Back plate kit for Versaflo™ and Speedglas™ series helmets
Catalogus # MMMAPZ3AF
Leverancier:   3M
Peltor™ Z3AF Back Plate
Catalogus # MMMAPZ3AF
Leverancier:  3M
Peltor™ Z3AF Back Plate
Peltor™ Z3AF Back Plate
Back plate kit for Versaflo™ and Speedglas™ series helmets
Price
17,70 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
This helmet assembly part securely attaches Peltor™ X-series earmuffs to Versaflo™ headtops M-series or Speedglas™ welding helmet 9100 MP, for effective hearing protection.
  • Attach earmuffs to a headtop or helmet
  • Check equipment regularly to increase service life
Ensure effective hearing protection when wearing a headtop or helmet with helmet assembly part Z3AF/2. This assembly part is designed to attach 3M™ Peltor™ X-series earmuffs to 3M™ Versaflo™ headtop M-series or 3M™ Speedglas™ welding helmet 9100 MP.
Specificaties
  • Omschrijving:
    Back plate kit for Versaflo™ and Speedglas™ series helmets