ContactUsLinkComponent
b.safe Air Valve UHPLC
b.safe Air Valve UHPLC
Catalog # BOHLM551-01
Supplier:  Bohlender
undefined
b.safe Air Valve UHPLC
Catalog # BOHLM551-01
Supplier:  Bohlender
Specifications
  • Omschrijving:
    Air valve UHPLC, service life 6 months, 2 per pack
  • VPE:
    1
Specifications
About this item
Valve body made of PP with connection (thread UNF ¹/₄" 28G) suitable for b.safe Caps.
  • Lid with ventilation opening and luerlock connection
  • Integrated check valve
  • Upstream membrane made of porous PTFE (0,1 μm)
  • Including attachable change indicator
For extraction of eluents. The b.safe Air Valve is especially adapted to the flow rate in UHPLC. The integrated non-return valve avoids evaporation of solvents and lets flow ambient air into the bottle in case of vacuum. The upstream porous PTFE membrane restrains pollutant particles coarse and fine dust, as well as microorganisms carried by it and other contaminations from outside.
The six months service life of the air valve can be easily monitored through either activating the display on the service life indicator and exchanging the valve in case of completely red colouring or documentation of the exchange date by hand and clip on the service life indicator at the valve.