Culti-Loops™ Lactobacillus paracasei subsp. paracasei ATCC™ BAA-52™
Catalog # OXOIR4609378
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- VPE:5
- Omschrijving:Lactobacillus paracasei subsp. paracasei ATCC™ BAA-52™
Specifications
About this item
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.