Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Staufen Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9877R-A680
 :  Bioss
Anti-Staufen Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9877R-A680
 :  Bioss

 

  • Type antilichaam:
    Primair
  • Clonality:
    Polyklonaal
  • Conjugatie:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunoFluorescentie:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Horse,
    Chicken,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitoop:
    421-520/577
  • Formulier:
    Liquid
  • Gen-ID:
    6780
  • Antigeen synoniemen:
    PPP1R150|STAU1|STAU|Double-stranded RNA-binding protein Staufen homolog 1
  • Modification:
    Unmodified
  • Opslagbuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Moleculair gewicht:
    63 kDa
  • Opslagtemperatuur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentratie:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogeen:
    KLH conjugated synthetic peptide derived from human Staufen
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Verpakking:
    Vial
  • VPE:
    100 µl

 

 

Binds double-stranded RNA (regardless of the sequence) and tubulin. May play a role in specific positioning of mRNAs at given sites in the cell by cross-linking cytoskeletal and RNA components, and in stimulating their translation at the site.

Type: Primary
Antigen: Staufen
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 421-520/577
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Horse, Others