Order Entry
Belgium
ContactUsLinkComponent
 
Anti-DOK1 Tyr362 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-5289R-A750
 :  Bioss
Anti-DOK1 Tyr362 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-5289R-A750
 :  Bioss

 

  • Type antilichaam:
    Primair
  • Clonality:
    Polyklonaal
  • Conjugatie:
    Alexa Fluor® 750
  • Flowcytometrie:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Formulier:
    Liquid
  • Gen-ID:
    1796
  • Antigeen synoniemen:
    DOK-1|p62dok|Docking protein 1 62kD|DOK1_HUMAN|Downstream of tyrosine kinase 1|DOK1 phospho Tyr362|P62DOK|DOK 1|docking protein 1 62kD downstream of tyrosine kinase 1|pp62|Docking protein 1|DOK1|DOK1 phospho Y362|p-DOK1 phospho Y362|docking protein 1 downstream of tyrosine kinase 1|Docking protein 1 p62dok Downstream of tyrosine kinase 1 pp62.
  • Modification:
    Phosphorylation (Tyr362)
  • Opslagbuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Moleculair gewicht:
    62 kDa
  • Opslagtemperatuur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentratie:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogeen:
    KLH conjugated synthetic phosphopeptide derived from human DOK1 around the phosphorylation site of Tyr362
  • Purification:
    Purified by Protein A
  • Verpakking:
    Vial
  • VPE:
    100 µl

 

 

Docking protein 1 is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukaemia (CML) patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. Docking protein 1 contains a putative pleckstrin homology domain at the amino terminus and ten PXXP SH3 recognition motifs. Docking protein 2 binds p120 (RasGAP) from CML cells. It has been postulated to play a role in mitogenic signaling.

Type: Primary
Antigen: DOK1 Tyr362
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human