Orders LinkContactUsLinkComponent
Sweeper
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.
Sweeper
Wisser 35 cm, 35 cm
Catalog # 129-0459
Supplier:   FHP VILEDA SCS
Sweeper
Catalog # 129-0459
Supplier:  FHP VILEDA SCS
Sweeper
Sweeper
Wisser 35 cm, 35 cm
Price
33,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The Sweeper and Pan is the perfect system for ergonomic and efficient dirt and liquid removal.
  • Flexible joint allows the user to reach into the most narrow spaces
  • Sweep up broken glass in your cleanroom quickly and easily
  • No need to change gloves when an item is dropped on the floor
  • Keeps workers from touching the floor within the cleanroom
  • Can be sprayed or wiped down with common disinfectants
  • Stands up easily for storage or can be hung
  • Adjustable bag holder for different size collection bags
The system is hygienic because of the washable and non-corroding material used in the construction. Overlapping rubber at the edges of the squeegee protect walls and furniture and the ergonomic grip reduces worker fatigue. Perfect for dry or damp conditions on smooth floors.
Ordering information: Sweeper and pan have to be ordered separately.
Specifications
  • Omschrijving:
    Wisser 35 cm
  • Seq:
    07
  • Breedte van de kop:
    35 cm