ContactUsLinkComponent
VWR®, Withdrawal System for Solvents with Manual Pressure Build-up for 10 and 30 L Stainless Steel Barrels
VWR®, Withdrawal System for Solvents with Manual Pressure Build-up for 10 and 30 L Stainless Steel Barrels
Catalog # 99900.001
Supplier:  VWR Chemicals
undefined
VWR®, Withdrawal System for Solvents with Manual Pressure Build-up for 10 and 30 L Stainless Steel Barrels
Catalog # 99900.001
Supplier:  VWR Chemicals
Specifications
  • Omschrijving:
    Withdrawal system for solvents with manual pressure build-up for 10 and 30 L stainless steel barrels
  • VPE:
    1
Specifications
About this item
Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 10 and 30 L stainless steel barrels.
  • Two dip tubes
  • Hand pump ball
  • Perfluorelastomer o-rings
This set contains a specially developed solvent withdrawal system for manual pressure build-up which can be used with VWR® 10 and 30 L returnable stainless steel barrels. This set includes two dip tubes, one for 10 L and one for 30 L barrels as well as a hand pump ball with a brass coupling.
Delivery: Complete set contains withdrawal system for 10 and 30 L barrels (stainless steel), Perfluorelastomer O-rings, 2 dip tubes (one for 10 L and one for 30 L), and hand pump ball with brass coupling.