Cold resistant gloves, EDGE® 48-193
Leverancier: Ansell
Over dit artikel
Cold-resistant safety gloves, offering mechanical protection for heavy-duty applications.
- PVC coating repels water, oils and greases, besides providing good dexterity and pliability at low temperatures
- Provides a barrier against cold, oils, greases and abrasions
- Durable polyester liner keeps hands warm and dry
Recommended for: Outdoor winter use, maintenance, storing and transportation.
Certificeringen: EN 388:2016 (3121A), EN 511 (011); Cat. II
2 Opties Hieronder Beschikbaar
Product Details & Documents
Cold-resistant safety gloves, offering mechanical protection for heavy-duty applications.
- PVC coating repels water, oils and greases, besides providing good dexterity and pliability at low temperatures
- Provides a barrier against cold, oils, greases and abrasions
- Durable polyester liner keeps hands warm and dry
Recommended for: Outdoor winter use, maintenance, storing and transportation.
Certificeringen: EN 388:2016 (3121A), EN 511 (011); Cat. II
Specifications for Product Family
Specificaties
- Catalog No:
- ANSE48-193/10
- ANSE48-193/11
- Ambidexter/handspecifiek:
- Handspecifiek
- Handspecifiek
- Voor eenmalig gebruik/herbruikbaar:
- Herbruikbaar
- Herbruikbaar
- Latex/latexvrij:
- Latexvrij
- Latexvrij
- Gepoederd/poedervrij:
- poedervrij
- poedervrij
- Voeringmateriaal:
- Polyester
- Polyester
- Afwerking:
- Volledig gecoat, PVC bumpers
- Volledig gecoat, PVC bumpers
- Manchetstijl:
- Veiligheidsmanchet
- Veiligheidsmanchet
- EN-norm:
- EN 388:2016 (3121A), EN 511 (011)
- EN 388:2016 (3121A), EN 511 (011)
- Sterility:
- Non sterile
- Non sterile
Specificaties
Product Family Options
Productinformatie
- HandschoenmaatKleurLengteBeschikbaarheidPrice
- Specificaties:
More
Specifications
Cat. No.ANSE48-193/10ModelAmbidexter/handspecifiekHandspecifiekVoor eenmalig gebruik/herbruikbaarHerbruikbaarLatex/latexvrijLatexvrijGepoederd/poedervrijpoedervrijVoeringmateriaalPolyesterAfwerkingVolledig gecoat, PVC bumpersManchetstijlVeiligheidsmanchetEN-normEN 388:2016 (3121A), EN 511 (011)SterilityNon sterile - Specificaties:
More
Specifications
Cat. No.ANSE48-193/11ModelAmbidexter/handspecifiekHandspecifiekVoor eenmalig gebruik/herbruikbaarHerbruikbaarLatex/latexvrijLatexvrijGepoederd/poedervrijpoedervrijVoeringmateriaalPolyesterAfwerkingVolledig gecoat, PVC bumpersManchetstijlVeiligheidsmanchetEN-normEN 388:2016 (3121A), EN 511 (011)SterilityNon sterile