Chemical resistant gloves, AlphaTec® 38-628
: Ansell
Combination of butyl and viton layer of 0,7 mm for a maximum protection against hazardous chemicals.
- Gloves are very flexible and provide excellent dexterity
- Designed for comfort, with a natural, curved ergonomic shape and soft feel
- Easy donning and good grip
- Absence of latex proteins means there is no risk of latex allergies
The 38-628 glove series offers superior protection against aliphatic, halogenated, and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids. These gloves provide the best resistance to the most aggressive chemicals, without compromising dexterity or comfort. Flexible and excellent dexterity, designed for comfort with a natural, curved ergonomic shape and soft feel.
Typical applications: Chemical handling; opening and draining pumps valves or lines; filling, blending and charging of raw materials; equipment repair & maintenance.
: EN ISO 374-1:2016 Type A (ABCDFLNS), EN ISO 374-5, EN 388:2016 (2120A), EN 1149; Cat. III
- Catalog No:
- 112-3819
- ANSE38-628/7
- 112-3820
- ANSE38-628/11
- ANSE38-628/8
- Ambidexter/handspecifiek:
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Voor eenmalig gebruik/herbruikbaar:
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Latex/latexvrij:
- Latexvrij
- Latexvrij
- Latexvrij
- Latexvrij
- Latexvrij
- Gepoederd/poedervrij:
- poedervrij
- poedervrij
- poedervrij
- poedervrij
- poedervrij
- Afwerking:
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Manchetstijl:
- Rolrand
- Rolrand
- Rolrand
- Rolrand
- Rolrand
- Handpalmdikte:
- 0,7 mm
- 0,7 mm
- 0,7 mm
- 0,7 mm
- 0,7 mm
- EN-norm:
- EN ISO 374-1:2016 Type A (ABCDFLNS), EN ISO 374-5, EN 388:2016 (2120A), EN 1149
- EN ISO 374-1:2016 Type A (ABCDFLNS), EN ISO 374-5, EN 388:2016 (2120A), EN 1149
- EN ISO 374-1:2016 Type A (ABCDFLNS), EN ISO 374-5, EN 388:2016 (2120A), EN 1149
- EN ISO 374-1:2016 Type A (ABCDFLNS), EN ISO 374-5, EN 388:2016 (2120A), EN 1149
- EN ISO 374-1:2016 Type A (ABCDFLNS), EN ISO 374-5, EN 388:2016 (2120A), EN 1149
- Antistatisch:
- +
- +
- +
- +
- +
Frequently Bought Together