TCBS agar
TCBS agar, 500 g
HIMEM189-500G
: HIMEDIA
TCBS agar, granulated is recommended for the selective isolation and cultivation of V. cholerae and other enteropathogenic Vibrio's causing food poisoning.
- VPE:500 g
- Omschrijving:TCBS agar
- Cat. No.:HIMEM189-500G
- Opslagtemperatuur:20...30 °C