Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Triamcinolone Hexacetonide, Reference Standard

Triamcinolone Hexacetonide, Reference Standard

Triamcinolone Hexacetonide, USP reference standard, Vial, 200 mg
Catalogus # 1679008.
Leverancier:   USP
Triamcinolone Hexacetonide, Reference Standard
Catalogus # 1679008.
Leverancier:  USP
CAS-nummer:  
Triamcinolone Hexacetonide, Reference Standard

Triamcinolone Hexacetonide, USP reference standard, Vial, 200 mg

CAS-nummer:  
Price
394,00 €
Log in voor beschikbaarheid
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certificeringen: US Pharmacopea Reference Standard

Specificaties

Specificaties

  • Verpakking:
    Vial
  • VPE:
    200 mg
  • Standaard:
    USP reference standard
  • Omschrijving:
    Triamcinolone Hexacetonide
  • Cat. No.:
    1679008.
  • Omschrijving:
    Triamcinolone Hexacetonide
  • Formule:
    C₃₀H₄₁FO₇
  • CAS No.:
    5611-51-8

Specificaties

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.