Orders LinkContactUsLinkComponent
Estradiol Cypionate System Suitability Mixture, Reference Standard
Estradiol Cypionate System Suitability Mixture, Reference Standard
Estradiol Cypionate System Suitability Mixture, USP reference standard, Vial, 10 mg
Catalogus # 1252193.
Leverancier:   USP
Estradiol Cypionate System Suitability Mixture, Reference Standard
Catalogus # 1252193.
Leverancier:  USP
CAS-nummer:  
Estradiol Cypionate System Suitability Mixture, Reference Standard
Estradiol Cypionate System Suitability Mixture, USP reference standard, Vial, 10 mg
CAS-nummer:  
Price
1 300,00 €
Log in voor beschikbaarheid
1 * 10 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
This mixture contains estradiol cypionate, estradiol-9-ene cypionate, estradiol dicypionate, and 4-methylestradiol cypionate
Certificeringen: US Pharmacopea Reference Standard
Specificaties
Specificaties
  • Verpakking:
    Vial
  • VPE:
    10 mg
  • Standaard:
    USP reference standard
  • Omschrijving:
    Estradiol Cypionate System Suitability Mixture
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.