Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Azithromycin Related Compounds, Reference Standard

Azithromycin Related Compounds, Reference Standard

Azithromycin Related Compound H, USP reference standard, Vial, 15 mg
Catalogus # 1046023.
Leverancier:   USP
Azithromycin Related Compounds, Reference Standard
Catalogus # 1046023.
Leverancier:  USP
CAS-nummer:  
Azithromycin Related Compounds, Reference Standard

Azithromycin Related Compound H, USP reference standard, Vial, 15 mg

CAS-nummer:  
Price
1 240,00 €
Log in voor beschikbaarheid
1 * 15 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certificeringen: US Pharmacopea Reference Standard

Specificaties

Specificaties

  • Verpakking:
    Vial
  • VPE:
    15 mg
  • Standaard:
    USP reference standard
  • Omschrijving:
    Azithromycin Related Compound H
  • Cat. No.:
    1046023.
  • Omschrijving:
    Azithromycin Related Compound H
  • Formule:
    C₄₅H₇₇N₃O₁₅S
  • Synonym(s):
    3'-N-{[4-(Acetylamino)phenyl]sulfonyl}-3'-N-demethylazithromycin
  • CAS No.:
    612069-30-4

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.